L(-)-Carnitin
Lieferant: Molekula
Synonyme:
Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
M59240682
MOLEM59240682EA
0
EUR
MOLEM59240682
L(-)-Carnitin
L(-)-Carnitin
Formel:
C₇H₁₅NO₃ Molecular Weight: 161,20 g/mol |
MDL:
MFCD00038747 CAS-Nummer: 541-15-1 EINECS: 208-768-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...