4-(2-Chlorethyl)morpholin Hydrochlorid

Lieferant: Molekula

Synonyme: N-(2-Chloroethyl)morpholine hydrochloride, N-(2-Chloroethyl)-Morpholinium Chloride

M60023687
MOLEM60023687EA 0 EUR
MOLEM60023687
4-(2-Chlorethyl)morpholin Hydrochlorid
4-(2-Chlorethyl)morpholin Hydrochlorid
Formel: C₆H₁₃Cl₂NO
Molecular Weight: 186,08 g/mol
MDL: MFCD00012797
CAS-Nummer: 3647-69-6
EINECS: 222-881-2
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR