Guanosin

Lieferant: Molekula

Synonyme: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

M60157874
MOLEM60157874EA 0 EUR
MOLEM60157874
Guanosin
Guanosin
Formel: C₁₀H₁₃N₅O₅
Molecular Weight: 283,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010182
CAS-Nummer: 118-00-3
EINECS: 204-227-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR