5-Mercapto-1-methyl-1H-tetrazol

Lieferant: Molekula

Synonyme: 5-Mercapto-1-methyltetrazole

M60268824
MOLEM60268824EA 0 EUR
MOLEM60268824
5-Mercapto-1-methyl-1H-tetrazol
5-Mercapto-1-methyl-1H-tetrazole
Formel: C₂H₄N₄S
Molecular Weight: 116,14 g/mol
MDL: MFCD00037317
CAS-Nummer: 13183-79-4
EINECS: 236-132-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR