(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Lieferant: Molekula

Synonyme: (R)-(+)-BINAP

M60347605
MOLEM60347605EA 0 EUR
MOLEM60347605
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formel: C₄₄H₃₂P₂
Molecular Weight: 622,69 g/mol
Schmelzpunkt: 239…242 °C
MDL: MFCD00010805
CAS-Nummer: 76189-55-4
Merck Index: 13,01223

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR