(±)-Campherchinon
Lieferant: Molekula
Synonyme:
1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone
M60570677
MOLEM60570677EA
0
EUR
MOLEM60570677
(±)-Campherchinon
(±)-Campherchinon
Formel:
C₁₀H₁₄O₂ Molecular Weight: 166.22 g/mol |
MDL:
MFCD00064160 CAS-Nummer: 10373-78-1 EINECS: 233-814-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...