Ethyl-3-methylcrotonat
Lieferant: Molekula
Synonyme:
3,3-Dimethylacrylic acid ethyl ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-methylbut-2-enoate, 3-Methyl-2-butenoic acid ethyl ester, 3-Methylcrotonic acid ethyl ester, Ethyl 3-Methyl-2-butenoate
M60762132
MOLEM60762132EA
0
EUR
MOLEM60762132
Ethyl-3-methylcrotonat
Ethyl-3-methylcrotonat
Formel:
C₇H₁₂O₂ Molecular Weight: 128,17 g/mol |
MDL:
MFCD00009187 CAS-Nummer: 638-10-8 EINECS: 211-319-1 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...