4-Cyan-2-nitroanilin

Lieferant: Molekula

Synonyme: 4-Cyano-2-nitroaniline

M60981240
MOLEM60981240EA 0 EUR
MOLEM60981240
4-Cyan-2-nitroanilin
4-Cyan-2-nitroanilin
Formel: C₇H₅N₃O₂
Molecular Weight: 163,14 g/mol
MDL: MFCD00013373
CAS-Nummer: 6393-40-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR