(R)-(-)-2-Amino-1-propanol

Lieferant: Molekula

Synonyme: D(-)-Alaninol, D(-)-2-Amino-1-propanol

M61082020
MOLEM61082020EA 0 EUR
MOLEM61082020
(R)-(-)-2-Amino-1-propanol
(R)-(-)-2-Amino-1-propanol
Formel: C₃H₉NO
Molecular Weight: 75,11 g/mol
Siedepunkt: 173 °C (1013 hPa)
Dichte: 0,96 g/cm³ (20 °C)
MDL: MFCD00064413
CAS-Nummer: 35320-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR