2-Chlor-5-methylpyridin
Lieferant: Molekula
|
Achtung
|
Synonyme:
2-Chloro-5-picoline
M61274719
MOLEM61274719EA
0
EUR
MOLEM61274719
2-Chlor-5-methylpyridin
2-Chlor-5-methylpyridin
Formel:
C₆H₆ClN Molecular Weight: 127,57 g/mol Siedepunkt: 97 °C (30 mmHg) Dichte: 1,169 g/cm³ (20 °C) |
MDL:
MFCD00792460 CAS-Nummer: 18368-64-4 EINECS: 418-050-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...