Thiazol-2-carbaldehyd

Lieferant: Molekula

Synonyme: 2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde

M61324267
MOLEM61324267EA 0 EUR
MOLEM61324267
Thiazol-2-carbaldehyd
Thiazol-2-carbaldehyd
Formel: C₄H₃NOS
Molecular Weight: 113,13 g/mol
MDL: MFCD00142924
CAS-Nummer: 10200-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR