2,5-Dimethylbenzoxazol

Lieferant: Molekula

M61658334
MOLEM61658334EA 0 EUR
MOLEM61658334
2,5-Dimethylbenzoxazol
2,5-Dimethylbenzoxazole
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
MDL: MFCD00005773
CAS-Nummer: 5676-58-4
EINECS: 227-136-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR