Bismarckbraun G

Lieferant: Molekula

M61849094
MOLEM61849094EA 0 EUR
MOLEM61849094
Bismarckbraun G
Bismarckbraun G
Formel: C₁₈H₁₈N8·2HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00012977
CAS-Nummer: 10114-58-6
EINECS: 233-314-3
Merck Index: 12,01294

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR