Amantadin

Lieferant: Molekula

Synonyme: 1-Aminotricyclo[3.3.1.1(3,7)]decan, 1-Adamantylamin, 1-Aminoadamantan

M61882414
MOLEM61882414EA 0 EUR
MOLEM61882414
Amantadin
Amantadin
Formel: C₁₀H₁₇N
Molecular Weight: 151,25 g/mol
Siedepunkt: 214 °C (1013 hPa)
Schmelzpunkt: 82 °C
MDL: MFCD00074732
CAS-Nummer: 768-94-5
EINECS: 212-201-2
Merck Index: 12,00389

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR