2-Hydroxynicotinsäure

Lieferant: Molekula

M61936762
MOLEM61936762EA 0 EUR
MOLEM61936762
2-Hydroxynicotinsäure
2-Hydroxynicotinsäure
Formel: C₆H₅NO₃
MDL: MFCD00010100
CAS-Nummer: 609-71-2
EINECS: 210-198-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR