2'-Desoxyadenosin-5'-triphosphat Dinatriumsalz (dATP Dinatriumsalz)

Lieferant: Molekula

Synonyme: 2'-Deoxyadenosine 5'-triphosphoric acid disodium salt, 2′-Deoxyadenosine 5′-triphosphate sodium salt, dATP-Na2

M61986848
MOLEM61986848EA 0 EUR
MOLEM61986848
2'-Desoxyadenosin-5'-triphosphat Dinatriumsalz (dATP Dinatriumsalz)
2'-Desoxyadenosin-5'-triphosphat Dinatriumsalz (dATP Dinatriumsalz)
Formel: C₁₀H₁₄N₅Na₂O₁₂P₃
Molecular Weight: 535,14 g/mol
MDL: MFCD00080335
CAS-Nummer: 74299-50-6
EINECS: 277-809-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR