2-Fluorphenylhydraziniumchlorid

Lieferant: Molekula

Synonyme: 2-Fluorphenylhydrazin Hydrochlorid, 1-(2-Fluorphenyl)hydrazin Hydrochlorid, 2-Fluorphenylhydrazinhydrochlorid

M62132853
MOLEM62132853EA 0 EUR
MOLEM62132853
2-Fluorphenylhydraziniumchlorid
2-Fluorphenylhydraziniumchlorid
Formel: C₆H₈ClFN₂
Molecular Weight: 162,59 g/mol
MDL: MFCD00012927
CAS-Nummer: 2924-15-4
EINECS: 220-885-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR