N,N-Diethyl-1,4-phenylendiammoniumsulfat
Lieferant: Molekula
|
Achtung
|
Synonyme:
N,N-Diethyl-p-phenylendiammoniumsulfat, N,N-Diethylbenzol-1,4-diammoniumsulfat
M62511832
MOLEM62511832EA
0
EUR
MOLEM62511832
N,N-Diethyl-1,4-phenylendiammoniumsulfat
N,N-Diethyl-1,4-phenylendiammoniumsulfat
Formel:
C₁₀H₁₈N₂O₄S Molecular Weight: 262,33 g/mol Siedepunkt: 274…275 °C (1013 hPa) Schmelzpunkt: 184…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012993 CAS-Nummer: 6283-63-2 EINECS: 228-500-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...