2-Brom-6-formylpyridin
Lieferant: Molekula
Synonyme:
6-Bromopyridine-2-carboxaldehyde, 6-Bromopyridine-2-carbaldehyde, 6-Bromo-2-pyridinecarboxaldehyde
M62552937
MOLEM62552937EA
0
EUR
MOLEM62552937
2-Brom-6-formylpyridin
2-Brom-6-formylpyridin
Formel:
C₆H₄BrNO Molecular Weight: 186,01 g/mol |
MDL:
MFCD02683546 CAS-Nummer: 34160-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...