1-Methylxanthin

Lieferant: Molekula

Synonyme: 1-Methyl-1H,3H-purin-2,6-dion, 3,7-Dihydro-1-methyl-1H-purin-2,6-dion, 1-Methyl-3,7-dihydro-2H-purin-2,6-dion, 2,6-Dihydroxy-1-methylpurin, 1-Methyl-2,6(1H,3H)-purindion, 1-Methyl-3,7-dihydro-1H-purin-2,6-dion

M62653874
MOLEM62653874EA 0 EUR
MOLEM62653874
1-Methylxanthin
1-Methylxanthin
Formel: C₆H₆N₄O₂
Molecular Weight: 166,14 g/mol
MDL: MFCD00005561
CAS-Nummer: 6136-37-4
EINECS: 228-108-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR