Polymyxin B-Sulfat

Lieferant: Molekula

M62768636
MOLEM62768636EA 0 EUR
MOLEM62768636
Polymyxin B-Sulfat
Polymyxin B sulphate
Formel: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MDL: MFCD00131991
CAS-Nummer: 1405-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR