Triethyl-2-phosphonopropionat

Lieferant: Molekula

Synonyme: Diethyl [1-(ethoxycarbonyl)ethyl]phosphonate, Ethyl 2-(diethoxyphosphoryl)propanoate

M62899313
MOLEM62899313EA 0 EUR
MOLEM62899313
Triethyl-2-phosphonopropionat
Triethyl-2-phosphonopropionat
Formel: C₉H₁₉O₅P
Molecular Weight: 238,22 g/mol
MDL: MFCD00009159
CAS-Nummer: 3699-66-9
EINECS: 223-033-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR