3-Acetyl-2,5-dichlorthiophen
Lieferant: Molekula
Synonyme:
1-(2,5-Dichlor-3-thienyl)-1-ethanon
M62999126
MOLEM62999126EA
0
EUR
MOLEM62999126
3-Acetyl-2,5-dichlorthiophen
3-Acetyl-2,5-dichlorthiophen
Formel:
C₆H₄Cl₂OS Molecular Weight: 195,07 g/mol Siedepunkt: 120…122 °C (6 mmHg) Schmelzpunkt: 37…40 °C |
MDL:
MFCD00014522 CAS-Nummer: 36157-40-1 EINECS: 252-893-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...