3-Hydroxy-2-nitropyridin

Lieferant: Molekula

M63119725
MOLEM63119725EA 0 EUR
MOLEM63119725
3-Hydroxy-2-nitropyridin
3-Hydroxy-2-nitropyridine
Formel: C₅H₄N₂O₃
Schmelzpunkt: 70 °C
MDL: MFCD00006258
CAS-Nummer: 15128-82-2
EINECS: 239-191-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR