2-Aminopyrimidin
Lieferant: Molekula
Synonyme:
Pyrimidin-2-ylamin
M63590011
MOLEM63590011EA
0
EUR
MOLEM63590011
2-Aminopyrimidin
2-Aminopyrimidin
Formel:
C₄H₅N₃ Molecular Weight: 95.1 g/mol Siedepunkt: 275 Schmelzpunkt: 123…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006089 CAS-Nummer: 109-12-6 EINECS: 203-648-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...