Boc-Pro-OH
Lieferant: Molekula
Synonyme:
N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
M63802560
MOLEM63802560EA
0
EUR
MOLEM63802560
Boc-Pro-OH
Boc-Pro-OH
Formel:
C₁₀H₁₇NO₄ Molecular Weight: 215.25 g/mol |
MDL:
MFCD00037324 CAS-Nummer: 15761-39-4 EINECS: 239-848-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...