2-Pyrazincarbonsäure
Lieferant: Molekula
Synonyme:
Pyrazincarbonsäure, Pyrazin-2-carbonsäure
M63860697
MOLEM63860697EA
0
EUR
MOLEM63860697
2-Pyrazincarbonsäure
Pyrazine-2-carboxylic acid
Formel:
C₅H₄N₂O₂ Molecular Weight: 124,1 g/mol Siedepunkt: 313 °C (1013 hPa) |
MDL:
MFCD00006130 CAS-Nummer: 98-97-5 EINECS: 202-718-1 Merck Index: 12,08143 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...