Fmoc-Pro-OH

Lieferant: Molekula

Synonyme: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline

M64210608
MOLEM64210608EA 0 EUR
MOLEM64210608
Fmoc-Pro-OH
Fmoc-Pro-OH
Formel: C₂₀H₁₉NO₄
Molecular Weight: 337,38 g/mol
MDL: MFCD00037122
CAS-Nummer: 71989-31-6
EINECS: 276-259-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR