(4-Hydroxyphenyl)acetamid

Lieferant: Molekula

M64453975
MOLEM64453975EA 0 EUR
MOLEM64453975
(4-Hydroxyphenyl)acetamid
(4-Hydroxyphenyl)acetamid
Formel: C₈H₉NO₂
MDL: MFCD00017145
CAS-Nummer: 17194-82-0
EINECS: 241-235-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR