2,3-Pyrazindicarbonsäure

Lieferant: Molekula

Synonyme: Pyrazin-2,3-dicarbonsäure

M64608623
MOLEM64608623EA 0 EUR
MOLEM64608623
2,3-Pyrazindicarbonsäure
Pyrazine-2,3-dicarboxylic acid
Formel: C₆H₄N₂O₄
Molecular Weight: 168.11 g/mol
MDL: MFCD00006131
CAS-Nummer: 89-01-0
EINECS: 201-875-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR