4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO freies Radikal)

Lieferant: Molekula

M64667867
MOLEM64667867EA 0 EUR
MOLEM64667867
4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO freies Radikal)
4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO)
Formel: C₉H₁₉N₂O
MDL: MFCD00006479
CAS-Nummer: 14691-88-4
EINECS: 238-738-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR