Salicylaldoxim

Lieferant: Molekula

Synonyme: 2-Hydroxybenzaldehyde oxime

M64864178
MOLEM64864178EA 0 EUR
MOLEM64864178
Salicylaldoxim
Salicylaldoxim
Formel: C₇H₇NO₂
MDL: MFCD00002120
CAS-Nummer: 94-67-7
EINECS: 202-353-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR