2-Hydroxycyclohexanon dimer

Lieferant: Molekula

M65156295
MOLEM65156295EA 0 EUR
MOLEM65156295
2-Hydroxycyclohexanon dimer
2-Hydroxycyclohexanon dimer
Formel: C₆H₁₀O₂
MDL: MFCD00151157
CAS-Nummer: 30282-14-5
UN: 1325
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR