2-Amino-5-chlorbenzophenon
Lieferant: Molekula
Synonyme:
1-Amino-4-chloro-2-(phenylmethanoyl)benzene, (2-Amino-5-chlorophenyl)(phenyl)methanone
M65620478
MOLEM65620478EA
0
EUR
MOLEM65620478
2-Amino-5-chlorbenzophenon
2-Amino-5-chlorbenzophenon
Formel:
C₁₃H₁₀ClNO Molecular Weight: 231,68 g/mol Siedepunkt: 424 °C (1013 hPa) Schmelzpunkt: 96…99 °C |
MDL:
MFCD00007839 CAS-Nummer: 719-59-5 EINECS: 211-949-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...