L(+)-Lysin Monohydrat

Lieferant: Molekula

Synonyme: K Monohydrat, (S)-(+)-2,6-Diaminohexansäure Monohydrat, H-Lys-OH.H2O, (S)-(+)-Lysin Monohydrat, (S)-(+)-2,6-Diaminocapronsäure Monohydrat, Lys Monohydrat

M66061013
MOLEM66061013EA 0 EUR
MOLEM66061013
L(+)-Lysin Monohydrat
L(+)-Lysin Monohydrat
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
Molecular Weight: 164,2 g/mol
Schmelzpunkt: 225 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00151035
CAS-Nummer: 39665-12-8
EINECS: 200-294-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR