Benzoin

Lieferant: Molekula

Synonyme: 2-Hydroxy-2-phenylacetophenone

M66437043
MOLEM66437043EA 0 EUR
MOLEM66437043
Benzoin
Benzoin
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
MDL: MFCD00004496
CAS-Nummer: 119-53-9
EINECS: 204-331-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR