7-Chlorchinaldin

Lieferant: Molekula

Synonyme: 7-Chlor-2-methylchinolin

M66515008
MOLEM66515008EA 0 EUR
MOLEM66515008
7-Chlorchinaldin
7-Chlorchinaldin
Formel: C₁₀H₈ClN
Molecular Weight: 177.63 g/mol
MDL: MFCD00075436
CAS-Nummer: 4965-33-7
EINECS: 440-600-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR