(Ethyl)triphenylphosphoniumbromid

Lieferant: Molekula
Achtung

Synonyme: Ethyltriphenylphosphonium bromide

M66589242
MOLEM66589242EA 0 EUR
MOLEM66589242
(Ethyl)triphenylphosphoniumbromid
(Ethyl)triphenylphosphoniumbromid
Formel: C₂₀H₂₀BrP
Molecular Weight: 371,26 g/mol
Schmelzpunkt: 205…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011838
CAS-Nummer: 1530-32-1
EINECS: 216-223-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR