Lanthan(III)trifluormethansulfonat Hydrat

Lieferant: Molekula

M66937880
MOLEM66937880EA 0 EUR
MOLEM66937880
Lanthan(III)trifluormethansulfonat Hydrat
Lanthan(III)trifluormethansulfonat Hydrat
Formel: C₃F₉LaO₉S₃·H₂O
CAS-Nummer: 34629-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR