Thiomorpholin

Lieferant: Molekula

M67046947
MOLEM67046947EA 0 EUR
MOLEM67046947
Thiomorpholin
Thiomorpholin
Formel: C₄H₉NS
Molecular Weight: 103,19 g/mol
MDL: MFCD00005974
CAS-Nummer: 123-90-0
EINECS: 204-660-2
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR