2-Phenylindol

Lieferant: Molekula

Synonyme: 2-Phenyl-1H-indole

M67315784
MOLEM67315784EA 0 EUR
MOLEM67315784
2-Phenylindol
2-Phenylindol
Formel: C₁₄H₁₁N
Molecular Weight: 193,25 g/mol
Siedepunkt: 420 °C (1013 hPa)
Schmelzpunkt: 186…187 °C
MDL: MFCD00005608
CAS-Nummer: 948-65-2
EINECS: 213-436-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR