1,2-Dimethylindol

Lieferant: Molekula

Synonyme: 1,2-Dimethyl-1H-indole, N,2-Dimethylindole

M67366623
MOLEM67366623EA 0 EUR
MOLEM67366623
1,2-Dimethylindol
1,2-Dimethylindol
Formel: C₁₀H₁₁N
Molecular Weight: 145,2 g/mol
MDL: MFCD00005802
CAS-Nummer: 875-79-6
EINECS: 212-877-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR