D(-)-Diisopropyltartrat
Lieferant: Molekula
Synonyme:
(2S,3S)-(-)-Diisopropyl tartrate, (2S,3S)-Diisopropyl 2,3-dihydroxysuccinate, D-(-)-Tartaric acid diisopropyl ester
M67988409
MOLEM67988409EA
0
EUR
MOLEM67988409
D(-)-Diisopropyltartrat
D(-)-Diisopropyltartrat
Formel:
C₁₀H₁₈O₆ Molecular Weight: 234,25 g/mol |
MDL:
MFCD00008876 CAS-Nummer: 62961-64-2 EINECS: 263-771-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...