3-Brombenzylamin Hydrochlorid

Lieferant: Molekula

Synonyme: (3-Bromophenyl)methanamine hydrochloride

M68181301
MOLEM68181301EA 0 EUR
MOLEM68181301
3-Brombenzylamin Hydrochlorid
3-Brombenzylamin Hydrochlorid
Formel: C₇H₉BrClN
Molecular Weight: 222,52 g/mol
MDL: MFCD00012856
CAS-Nummer: 39959-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR