3-Bromthioanisol

Lieferant: Molekula

Synonyme: 1-Brom-3-(methylthio)benzol, 3-Bromphenylmethylsulfid

M68453130
MOLEM68453130EA 0 EUR
MOLEM68453130
3-Bromthioanisol
3-Bromthioanisol
Formel: C₇H₇BrS
Molecular Weight: 203,1 g/mol
MDL: MFCD00041395
CAS-Nummer: 33733-73-2
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR