Cisplatin
Lieferant: Molekula
Synonyme:
cis-Diammindichloridoplatin(II), Platinol
M68655191
MOLEM68655191EA
0
EUR
MOLEM68655191
Cisplatin
cis-Platin
Formel:
H₆Cl₂N₂Pt Molecular Weight: 300,05 g/mol Dichte: 3,738 g/cm³ (20 °C) |
MDL:
MFCD00011623 CAS-Nummer: 15663-27-1 EINECS: 239-733-8 UN: 3288 ADR: 6.1,II Merck Index: 13,02341 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...