6-Nitro-m-kresol
Lieferant: Molekula
|
Gefahr
|
Synonyme:
1-Hydroxy-3-methyl-6-nitrobenzol, 3-Hydroxy-4-nitrotoluol, 5-Methyl-2-nitrophenol
M69284514
MOLEM69284514EA
0
EUR
MOLEM69284514
6-Nitro-m-kresol
6-Nitro-m-kresol
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Schmelzpunkt: 53…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007111 CAS-Nummer: 700-38-9 EINECS: 211-843-0 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...