1-Naphthylphosphat Mononatriumsalz Monohydrat

Lieferant: Molekula
Achtung

Synonyme: 1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate

M69305565
MOLEM69305565EA 0 EUR
MOLEM69305565
1-Naphthylphosphat Mononatriumsalz Monohydrat
1-Naphthylphosphat Mononatriumsalz Monohydrat
Formel: C₁₀H₈NaO₄P·1H₂O
Molecular Weight: 264,15 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00150615
CAS-Nummer: 81012-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR