1-(4-Nitrophenyl)glycerin
Lieferant: Molekula
Synonyme:
1-(4-Nitrophenyl)-1,2,3-propantriol, PNPG
M69318582
MOLEM69318582EA
0
EUR
MOLEM69318582
1-(4-Nitrophenyl)glycerin
1-(4-Nitrophenyl)glycerin
Formel:
C₉H₁₁NO₅ Molecular Weight: 213,19 g/mol Schmelzpunkt: 94…97 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00017037 CAS-Nummer: 2207-68-3 EINECS: 218-624-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...