1-Acetyl-2-phenylhydrazin
Lieferant: Molekula
Synonyme:
2-Phenylacetohydrazide, N-Acetyl-N'-phenylhydrazine
M69389939
MOLEM69389939EA
0
EUR
MOLEM69389939
1-Acetyl-2-phenylhydrazin
1-Acetyl-2-phenylhydrazin
Formel:
C₈H₁₀N₂O Molecular Weight: 150,18 g/mol |
MDL:
MFCD00008672 CAS-Nummer: 114-83-0 EINECS: 204-055-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...